MAT047029705 Matrix Scientific

5-Chloro-2-cyanopyridine

  • MDL number MFCD03788835
  • Cas Number : 89809-64-3
  • Molecular Weight : 138.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83-84
Purity 95
Country of Origin USA
Boiling point 110�Torr
Product Link http://www.matrixscientific.com/011186.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011186.html
Availability details for
Quantity
  • Availability
  • Add to Cart