MAT047029721 Matrix Scientific

1-(2,6-Dimethoxybenzyl)piperidin-4-ylaminedihydrochloride

  • MDL number MFCD06247029
  • Cas Number : 849925-08-2
  • Molecular Weight : 323.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011220.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011220.html
Availability details for
Quantity
  • Availability
  • Add to Cart