MAT047029722 Matrix Scientific

4,5,6-Trifluoropyrimidine

  • MDL number MFCD05662363
  • Cas Number : 17573-78-3
  • Molecular Weight : 134.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Ambient
Country of Origin USA
Boiling point 83-85�0mm
Product Link http://www.matrixscientific.com/011226.html

Safety Information

hcodes Flammable
MSDS http://www.matrixscientific.com/011226.html
Availability details for
Quantity
  • Availability
  • Add to Cart