MAT047029743 Matrix Scientific

Benzo[1,3]dioxol-5-ylmethyl-(1-methyl-piperidin-4-yl)-amine

  • MDL number MFCD02051542
  • Cas Number : 416865-72-0
  • Molecular Weight : 248.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010795.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010795.html
Availability details for
Quantity
  • Availability
  • Add to Cart