MAT047029744 Matrix Scientific

(1-Methyl-piperidin-4-yl)-(tetrahydro-furan-2-yl-methyl)-amine

  • MDL number MFCD01454093
  • Cas Number : 416869-66-4
  • Molecular Weight : 198.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010796.html

Safety Information

MSDS http://www.matrixscientific.com/010796.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart