MAT047029747 Matrix Scientific

Pyridine-2-carboxylic acid [3-(3-dimethylamino-propylamino)-propyl]-amide

  • MDL number MFCD03682019
  • Cas Number : 510764-56-4
  • Molecular Weight : 264.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010799.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010799.html
Availability details for
Quantity
  • Availability
  • Add to Cart