MAT047029761 Matrix Scientific

Cyclohex-3-enylmethyl-(3-imidazol-1-yl-propyl)-amine

  • MDL number MFCD03724740
  • Cas Number : 626209-23-2
  • Molecular Weight : 219.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010812.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010812.html
Availability details for
Quantity
  • Availability
  • Add to Cart