MAT047029767 Matrix Scientific

(2-Fluoro-benzyl)-(3-imidazol-1-yl-propyl)-amine

  • MDL number MFCD04035527
  • Cas Number : 626209-31-2
  • Molecular Weight : 233.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010817.html

Safety Information

MSDS http://www.matrixscientific.com/010817.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart