MAT047029794 Matrix Scientific

Cyclopentyl-(2-fluoro-benzyl)-amine

  • MDL number MFCD04035530
  • Cas Number : 85952-80-3
  • Molecular Weight : 193.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010841.html

Safety Information

MSDS http://www.matrixscientific.com/010841.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart