MAT047029803 Matrix Scientific

Cyclopropyl-thiophen-2-ylmethyl-amine

  • MDL number MFCD03856576
  • Cas Number : 14471-18-2
  • Molecular Weight : 153.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-80
Country of Origin USA
Boiling point 78-90�Torr
Product Link http://www.matrixscientific.com/010849.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010849.html
Availability details for
Quantity
  • Availability
  • Add to Cart