MAT047029808 Matrix Scientific

6'-Chloro-2'-fluoro-3'-methylacetophenone

  • MDL number MFCD01631378
  • Cas Number : 261762-78-1
  • Molecular Weight : 186.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/002047.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002047.html
Availability details for
Quantity
  • Availability
  • Add to Cart