MAT047029844 Matrix Scientific

4-Chloro-6-morpholin-4-yl-pyrimidin-5-ylamine

  • MDL number MFCD00473322
  • Cas Number : 84762-69-6
  • Molecular Weight : 214.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010886.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010886.html
Availability details for
Quantity
  • Availability
  • Add to Cart