MAT047029852 Matrix Scientific

6-Chloro-2-fluoro-3-methylbenzoic acid

  • MDL number MFCD01631369
  • Cas Number : 32890-90-7
  • Molecular Weight : 188.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/002051.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002051.html
Availability details for
Quantity
  • Availability
  • Add to Cart