MAT047029865 Matrix Scientific

(5-Methyl-furan-2-ylmethyl)-(2,2,6,6-tetramethyl-piperidin-4-yl)-amine

  • MDL number MFCD03725048
  • Cas Number : 626213-21-6
  • Molecular Weight : 250.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010905.html

Safety Information

MSDS http://www.matrixscientific.com/010905.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart