MAT047029867 Matrix Scientific

Cyclopentyl-(5-methyl-furan-2-ylmethyl)-amine

  • MDL number MFCD00972782
  • Cas Number : 142920-61-4
  • Molecular Weight : 179.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 120-121� Torr
Product Link http://www.matrixscientific.com/010907.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010907.html
Availability details for
Quantity
  • Availability
  • Add to Cart