MAT047029874 Matrix Scientific

6-Chloro-2-fluoro-3-methylbenzyl alcohol

  • MDL number MFCD01631372
  • Cas Number : 261762-84-9
  • Molecular Weight : 174.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56-58
Country of Origin USA
Product Link http://www.matrixscientific.com/002053.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002053.html
Availability details for
Quantity
  • Availability
  • Add to Cart