MAT047029882 Matrix Scientific

Cyclopentyl-(1-phenyl-ethyl)-amine

  • MDL number MFCD03640281
  • Cas Number : 626213-92-1
  • Molecular Weight : 189.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010921.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010921.html
Availability details for
Quantity
  • Availability
  • Add to Cart