MAT047029888 Matrix Scientific

3-(1-Pyridin-2-yl-ethylamino)-propan-1-ol

  • MDL number MFCD04032682
  • Cas Number : 511237-61-9
  • Molecular Weight : 180.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010925.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010925.html
Availability details for
Quantity
  • Availability
  • Add to Cart