MAT047029894 Matrix Scientific

3-[(Furan-2-ylmethyl)-amino]-propan-1-ol

  • MDL number MFCD03725083
  • Cas Number : 137788-52-4
  • Molecular Weight : 155.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.0681/20
Country of Origin USA
Boiling point 142-143� Torr
Product Link http://www.matrixscientific.com/010931.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010931.html
Availability details for
Quantity
  • Availability
  • Add to Cart