MAT047029896 Matrix Scientific

(3-Methoxy-propyl)-(1-pyridin-2-yl-ethyl)-amine

  • MDL number MFCD03821897
  • Cas Number : 626214-26-4
  • Molecular Weight : 194.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010933.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010933.html
Availability details for
Quantity
  • Availability
  • Add to Cart