MAT047029912 Matrix Scientific

1-(4-Methyl-pyridin-2-yl)-1H-pyrrole-2-carbaldehyde

  • MDL number MFCD02665159
  • Cas Number : 383136-14-9
  • Molecular Weight : 186.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010947.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010947.html
Availability details for
Quantity
  • Availability
  • Add to Cart