MAT047029917 Matrix Scientific

Isopropyl-(tetrahydro-furan-2-ylmethyl)-amine

  • MDL number MFCD03856628
  • Cas Number : 5064-46-0
  • Molecular Weight : 143.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 68� Torr
Product Link http://www.matrixscientific.com/010952.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010952.html
Availability details for
Quantity
  • Availability
  • Add to Cart