MAT047029921 Matrix Scientific

2-Isobutyl-3-oxo-2,3-dihydro-1H-isoindole-4-carboxylic acid

  • MDL number MFCD03821910
  • Cas Number : 626215-95-0
  • Molecular Weight : 233.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010955.html

Safety Information

MSDS http://www.matrixscientific.com/010955.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart