MAT047029936 Matrix Scientific

[2-(3,4-Dimethoxy-phenyl)-ethyl]-pyridin-2-yl-methyl-amine

  • MDL number MFCD01135628
  • Cas Number : 99615-36-8
  • Molecular Weight : 272.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010969.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010969.html
Availability details for
Quantity
  • Availability
  • Add to Cart