MAT047029950 Matrix Scientific

N'-(2,3-Dimethoxy-benzyl)-N,N-dimethyl-ethane-1,2-diamine

  • MDL number MFCD03856642
  • Cas Number : 626216-30-6
  • Molecular Weight : 238.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010982.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010982.html
Availability details for
Quantity
  • Availability
  • Add to Cart