MAT047029957 Matrix Scientific

N'-Furan-2-ylmethyl-N,N-dimethyl-ethane-1,2-diamine

  • MDL number MFCD03856648
  • Cas Number : 626216-39-5
  • Molecular Weight : 168.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 165-175� Torr
Product Link http://www.matrixscientific.com/010988.html

Safety Information

MSDS http://www.matrixscientific.com/010988.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart