MAT047029960 Matrix Scientific

N'-(4-Fluoro-benzyl)-N,N-dimethyl-propane-1,3-diamine

  • MDL number MFCD03856651
  • Cas Number : 626216-48-6
  • Molecular Weight : 210.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010991.html

Safety Information

MSDS http://www.matrixscientific.com/010991.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart