MAT047029967 Matrix Scientific

N,N-Dimethyl-N'-pyridin-3-ylmethyl-propane-1,3-diamine

  • MDL number MFCD03856655
  • Cas Number : 136469-88-0
  • Molecular Weight : 193.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010997.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010997.html
Availability details for
Quantity
  • Availability
  • Add to Cart