MAT047029979 Matrix Scientific

(1-Methyl-piperidin-4-yl)-pyridin-3-ylmethyl-amine

  • MDL number MFCD03856660
  • Cas Number : 626217-79-6
  • Molecular Weight : 205.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011008.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011008.html
Availability details for
Quantity
  • Availability
  • Add to Cart