MAT047029986 Matrix Scientific

N,N-Diethyl-N'-(1-ethyl-piperidin-4-yl)-ethane-1,2-diamine

  • MDL number MFCD04035539
  • Cas Number : 626217-94-5
  • Molecular Weight : 227.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011014.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011014.html
Availability details for
Quantity
  • Availability
  • Add to Cart