MAT047029989 Matrix Scientific

(3-Cyano-indol-1-yl)-acetic acid

  • MDL number MFCD03306285
  • Cas Number : 626218-03-9
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011017.html

Safety Information

MSDS http://www.matrixscientific.com/011017.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart