MAT047030000 Matrix Scientific

4-(4-Benzylpiperazino)aniline

  • MDL number MFCD00172704
  • Cas Number : 16154-69-1
  • Molecular Weight : 267.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136
Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/011026.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011026.html
Availability details for
Quantity
  • Availability
  • Add to Cart