MAT047030001 Matrix Scientific

(3-Furan-2-yl-1-methyl-propyl)-methyl-amine

  • MDL number MFCD00520659
  • Cas Number : 1593-37-9
  • Molecular Weight : 153.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 115� Torr
Product Link http://www.matrixscientific.com/011027.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011027.html
Availability details for
Quantity
  • Availability
  • Add to Cart