MAT047030003 Matrix Scientific

8-Chloro-4-furan-2-yl-2,3,3a,4,5,9b-hexa-hydrofuro[3,2-c]quinoline

  • MDL number MFCD03856666
  • Cas Number : 1164485-17-9
  • Molecular Weight : 275.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011029.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011029.html
Availability details for
Quantity
  • Availability
  • Add to Cart