MAT047030007 Matrix Scientific

5-Methyl-7-phenyl-4,5,6,7-tetrahydro-[1,2,4]triazolo[1,5-a]pyrimidine

  • MDL number MFCD01627098
  • Cas Number : 133533-72-9
  • Molecular Weight : 214.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011033.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011033.html
Availability details for
Quantity
  • Availability
  • Add to Cart