MAT047030009 Matrix Scientific

7-Furan-2-yl-5-phenyl-4,5,6,7-tetrahydro-[1,2,4]triazolo[1,5-a]pyrimidine

  • MDL number MFCD00613798
  • Cas Number : 301320-67-2
  • Molecular Weight : 266.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011034.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011034.html
Availability details for
Quantity
  • Availability
  • Add to Cart