MAT047030011 Matrix Scientific

4-Oxo-3-thiophen-2-ylmethyl-10-oxa-3-aza-tricyclo-[5.2.1.0*1,5*]dec-8-ene-6-carboxylic acid

  • MDL number MFCD04116079
  • Cas Number : 1185549-47-6
  • Molecular Weight : 291.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011036.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011036.html
Availability details for
Quantity
  • Availability
  • Add to Cart