MAT047030018 Matrix Scientific

2-Pyridin-4-yl-benzooxazol-5-ylamine

  • MDL number MFCD00448006
  • Cas Number : 349609-85-4
  • Molecular Weight : 211.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/011043.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011043.html
Availability details for
Quantity
  • Availability
  • Add to Cart