MAT047030045 Matrix Scientific

(2-Methyl-imidazol-1-yl)-acetic acid

  • MDL number MFCD03250177
  • Cas Number : 25023-36-3
  • Molecular Weight : 140.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011067.html

Safety Information

MSDS http://www.matrixscientific.com/011067.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart