MAT047030061 Matrix Scientific

2-Amino-5-cyanobenzotrifluoride

  • MDL number MFCD00275473
  • Cas Number : 327-74-2
  • Molecular Weight : 186.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64-65
Purity 98
Country of Origin USA
Boiling point 108-110�Torr
Product Link http://www.matrixscientific.com/011083.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/011083.html
Availability details for
Quantity
  • Availability
  • Add to Cart