MAT047030123 Matrix Scientific

2-Bromo-5-cyanopyridine

  • MDL number MFCD00234141
  • Cas Number : 139585-70-9
  • Molecular Weight : 183.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116-118
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/011143.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011143.html
Availability details for
Quantity
  • Availability
  • Add to Cart