MAT047030131 Matrix Scientific

2-(6-Amino-9H-purin-8-ylsulfanyl)-butyric acid

  • MDL number MFCD03446833
  • Cas Number : 436086-77-0
  • Molecular Weight : 253.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010755.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010755.html
Availability details for
Quantity
  • Availability
  • Add to Cart