MAT047030172 Matrix Scientific

(4-Methoxy-benzyl)-(3-morpholin-4-yl-propyl)-amine

  • MDL number MFCD01475412
  • Cas Number : 436096-93-4
  • Molecular Weight : 264.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010763.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010763.html
Availability details for
Quantity
  • Availability
  • Add to Cart