MAT047030179 Matrix Scientific

(4-Chloro-benzyl)-(3-morpholin-4-yl-propyl)-amine

  • MDL number MFCD01475415
  • Cas Number : 107921-37-9
  • Molecular Weight : 268.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 207-209�Torr
Product Link http://www.matrixscientific.com/010765.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010765.html
Availability details for
Quantity
  • Availability
  • Add to Cart