MAT047030183 Matrix Scientific

(3-Methoxy-benzyl)-(3-morpholin-4-yl-propyl)-amine

  • MDL number MFCD03724728
  • Cas Number : 57095-67-7
  • Molecular Weight : 264.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010769.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010769.html
Availability details for
Quantity
  • Availability
  • Add to Cart