MAT047030195 Matrix Scientific

4-Iodophenylacetic acid

  • MDL number MFCD00082985
  • Cas Number : 1798-06-7
  • Molecular Weight : 262.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-136
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/011210.html

Safety Information

hcodes Irritant/Light Sensitive
MSDS http://www.matrixscientific.com/011210.html
Availability details for
Quantity
  • Availability
  • Add to Cart