MAT047030234 Matrix Scientific

(5-Methyl-furan-2-ylmethyl)-pyridin-3-ylmethyl-amine

  • MDL number MFCD03724730
  • Cas Number : 510764-54-2
  • Molecular Weight : 202.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010779.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010779.html
Availability details for
Quantity
  • Availability
  • Add to Cart