MAT047030239 Matrix Scientific

2-Methoxy-5-pyridinecarboxylic acid

  • MDL number MFCD04115721
  • Cas Number : 66572-55-2
  • Molecular Weight : 153.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171-172
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/011264.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011264.html
Availability details for
Quantity
  • Availability
  • Add to Cart