MAT047030258 Matrix Scientific

2-Bromo-5-fluorobenzyl bromide

  • MDL number MFCD00042187
  • Cas Number : 112399-50-5
  • Molecular Weight : 267.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 33-36
Purity 98
Country of Origin USA
Boiling point 89-90�m
Product Link http://www.matrixscientific.com/011284.html

Safety Information

hcodes Corrosive/Refrigerate/Lachrymator
MSDS http://www.matrixscientific.com/011284.html
Availability details for
Quantity
  • Availability
  • Add to Cart