MAT047030273 Matrix Scientific

1-Ethyl-2-methyl-1H-benzoimidazole-5-carboxylicacid ethyl ester

  • MDL number MFCD00447373
  • Cas Number : 92108-02-6
  • Molecular Weight : 232.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87
Country of Origin USA
Product Link http://www.matrixscientific.com/011298.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011298.html
Availability details for
Quantity
  • Availability
  • Add to Cart